siRNA Details
Gene Symbol: Olfr545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146840.1 | NP_667051.1 | 1 | 610 | ||
GenBank mRNA | BC106987.1 | - | 1 | 610 | 40043277 | |
BC128265.1 | - | 1 | 610 | 40043266 | ||
BC128266.1 | - | 1 | 610 | 40043268 |
