siRNA Details
| Gene Symbol: Olfr1239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146970.1 | NP_667181.1 | 1 | 330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1239| Gene Symbol: Olfr1239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146970.1 | NP_667181.1 | 1 | 330 | ||