siRNA Details
| Gene Symbol: Olfr1052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147010.2 | NP_667221.2 | 1 | 487 | ||
| GenBank mRNA | BC127979.1 | - | 1 | 491 | 40135213 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1052| Gene Symbol: Olfr1052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147010.2 | NP_667221.2 | 1 | 487 | ||
| GenBank mRNA | BC127979.1 | - | 1 | 491 | 40135213 | |