siRNA Details
| Gene Symbol: Olfr711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147035.2 | NP_667246.2 | 3 | 1274 | ||
| GenBank mRNA | BC132382.1 | - | 1 | 945 | 40130659 | |
| BC138433.1 | - | 1 | 945 | 8861453 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr711| Gene Symbol: Olfr711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147035.2 | NP_667246.2 | 3 | 1274 | ||
| GenBank mRNA | BC132382.1 | - | 1 | 945 | 40130659 | |
| BC138433.1 | - | 1 | 945 | 8861453 | ||