siRNA Details
Gene Symbol: Olfr283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147036.1 | NP_667247.1 | 1 | 907 | ||
GenBank mRNA | BC125358.1 | - | 1 | 996 | 40129873 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147036.1 | NP_667247.1 | 1 | 907 | ||
GenBank mRNA | BC125358.1 | - | 1 | 996 | 40129873 |