siRNA Details
| Gene Symbol: Olfr652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147048.2 | NP_667259.2 | 1 | 504 | ||
| GenBank mRNA | BC137763.1 | - | 1 | 504 | 8860782 | |
| BC137764.1 | - | 1 | 504 | 8860783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr652| Gene Symbol: Olfr652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147048.2 | NP_667259.2 | 1 | 504 | ||
| GenBank mRNA | BC137763.1 | - | 1 | 504 | 8860782 | |
| BC137764.1 | - | 1 | 504 | 8860783 | ||