siRNA Details
| Gene Symbol: Olfr603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147070.2 | NP_667281.2 | 1 | 278 | ||
| Gene Symbol: Olfr596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190381.1 | NP_001177310.1 | 1 | 278 | ||

Olfr603
Genomic Map