siRNA Details
| Gene Symbol: Olfr619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147076.2 | NP_667287.2 | 1 | 320 | ||
| GenBank mRNA | BC141053.1 | - | 1 | 320 | 9055665 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr619| Gene Symbol: Olfr619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147076.2 | NP_667287.2 | 1 | 320 | ||
| GenBank mRNA | BC141053.1 | - | 1 | 320 | 9055665 | |