siRNA Details
Gene Symbol: Olfr619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147076.2 | NP_667287.2 | 1 | 720 | ||
GenBank mRNA | BC141053.1 | - | 1 | 720 | 9055665 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147076.2 | NP_667287.2 | 1 | 720 | ||
GenBank mRNA | BC141053.1 | - | 1 | 720 | 9055665 |