siRNA Details
Gene Symbol: Olfr549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147101.2 | NP_667312.2 | 1 | 917 | ||
GenBank mRNA | BC100691.1 | - | 1 | 916 | ||
BC100692.1 | - | 1 | 916 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147101.2 | NP_667312.2 | 1 | 917 | ||
GenBank mRNA | BC100691.1 | - | 1 | 916 | ||
BC100692.1 | - | 1 | 916 |