siRNA Details
| Gene Symbol: Olfr577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147109.1 | NP_667320.1 | 1 | 282 | ||
| GenBank mRNA | BC104082.1 | - | 1 | 281 | ||
| BC104083.1 | - | 1 | 281 | |||
| BC119411.1 | - | 1 | 282 | 40042081 | ||
| BC119412.2 | - | 1 | 282 | |||

Olfr577
Genomic Map