siRNA Details
| Gene Symbol: Olfr575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147114.2 | NP_667325.1 | 1 | 840 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr575| Gene Symbol: Olfr575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147114.2 | NP_667325.1 | 1 | 840 | ||