siRNA Details
| Gene Symbol: Olfr632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147119.1 | NP_667330.1 | 1 | 383 | ||
| GenBank mRNA | BC106823.2 | - | 1 | 383 | 40043471 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr632| Gene Symbol: Olfr632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147119.1 | NP_667330.1 | 1 | 383 | ||
| GenBank mRNA | BC106823.2 | - | 1 | 383 | 40043471 | |