siRNA Details
| Gene Symbol: Phldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252442.1 | NP_001239371.1 | 18 | 3880 | ||
| NM_153412.4 | NP_700461.2 | 17 | 3721 | |||
| GenBank mRNA | AF506821.1 | - | 17 | 3722 | ||
| AK035655.1 | - | 12 | 1484 | |||
| AK087133.1 | - | 2 | 109 | |||
| AK202362.1 | - | 2 | 44 | |||
| BC050915.1 | - | 18 | 3736 | 6405379 | ||
| BC060683.1 | - | 18 | 3736 | 6405379 | ||

Phldb2
Genomic Map