siRNA Details
| Gene Symbol: Lcor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172154.4 | NP_742166.1 | 1 | 2940 | ||
| GenBank mRNA | AB076079.1 | - | 6 | 551 | ||
| AK041090.1 | - | 1 | 2941 | |||
| AK041621.1 | - | 6 | 754 | |||
| AK077424.1 | - | 6 | 586 | |||
| AK129442.1 | - | 4 | 376 | |||
| AK155007.1 | - | 6 | 630 | |||
| BC090652.1 | - | 3 | 334 | 30657664 | ||
| BC099938.1 | - | 6 | 622 | 30085146 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008760473.

Lcor
Genomic Map