siRNA Details
| Gene Symbol: Snx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172277.2 | NP_758481.1 | 6 | 668 | ||
| GenBank mRNA | AK045061.1 | - | 6 | 661 | ||
| AK148429.1 | - | 6 | 661 | |||
| AK156817.1 | - | 6 | 614 | |||
| AK161672.1 | - | 5 | 602 | |||
| BC019142.1 | - | 5 | 664 | |||
| BC037599.1 | - | 6 | 694 | 5150754 | ||

Snx8
Genomic Map