siRNA Details
| Gene Symbol: Snx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172277.2 | NP_758481.1 | 11 | 1285 | ||
| GenBank mRNA | AK045061.1 | - | 11 | 1278 | ||
| AK082871.1 | - | 11 | 1278 | |||
| AK148429.1 | - | 11 | 1278 | |||
| AK156817.1 | - | 11 | 1231 | |||
| AK161672.1 | - | 10 | 1219 | |||
| BC019142.1 | - | 10 | 1281 | |||
| BC037599.1 | - | 11 | 1311 | 5150754 | ||

Snx8
Genomic Map