siRNA Details
| Gene Symbol: Zfp536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172385.2 | NP_759017.1 | 4 | 1528 | ||
| GenBank mRNA | AK030633.1 | - | 4 | 1529 | ||
| AK034508.1 | - | 4 | 1472 | |||
| AK122270.1 | - | 5 | 1535 | |||
| BC033594.1 | - | 4 | 1450 | 5367892 | ||

Zfp536
Genomic Map