siRNA Details
| Gene Symbol: Slc25a32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172402.3 | NP_765990.2 | 6 | 896 | ||
| GenBank mRNA | AK031187.1 | - | 6 | 701 | ||
| AK145178.1 | - | 6 | 856 | |||
| AK167501.1 | - | 6 | 706 | |||
| AK170643.1 | - | 6 | 865 | |||
| AK172029.1 | - | 6 | 864 | |||
| AK172597.1 | - | 6 | 809 | |||
| BC031874.1 | - | 6 | 830 | 5149984 | ||

Slc25a32
Genomic Map