siRNA Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172410.2 | NP_765998.1 | 12 | 1338 | ||
| GenBank mRNA | AK010450.1 | - | 11 | 1289 | ||
| AK028864.1 | - | 12 | 1339 | |||
| AK045380.1 | - | 1197 | ||||
| AK129055.1 | - | 11 | 1288 | |||
| AK167105.1 | - | 11 | 1289 | |||
| AK169194.1 | - | 9 | 1059 | |||
| AK169693.1 | - | 11 | 1308 | |||
| BC023140.1 | - | 11 | 1284 | 3991335 | ||

Nup93
Genomic Map