siRNA Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159527.1 | NP_001152999.1 | 9 | 1100 | ||
| NM_172470.3 | NP_766058.3 | 9 | 1100 | |||
| GenBank mRNA | AK015490.1 | - | 9 | 1065 | ||
| AK029896.1 | - | 9 | 1055 | |||
| AK031061.1 | - | 9 | 1091 | |||
| AK053724.1 | - | 8 | 921 | |||
| AK083971.1 | - | 9 | 1045 | |||
| AK173151.1 | - | 9 | 1042 | |||
| BC056925.1 | - | 9 | 1209 | 5704780 | ||

Wdr35
Genomic Map