siRNA Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172961.3 | NP_766549.2 | 13 | 1300 | ||
| GenBank mRNA | AK132282.1 | - | 13 | 1249 | ||
| AK165348.1 | - | 13 | 1251 | |||
| BC058079.1 | - | 13 | 1240 | 6826741 | ||
| BC058521.1 | - | 13 | 1240 | 6825487 | ||

Abat
Genomic Map