siRNA Details
| Gene Symbol: Atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174874.3 | NP_777363.1 | 13 | 1788 | ||
| GenBank mRNA | AK028712.1 | - | 13 | 1799 | ||
| AK031570.1 | - | 13 | 1789 | |||
| AK144239.1 | - | 13 | 1793 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atg4b| Gene Symbol: Atg4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174874.3 | NP_777363.1 | 13 | 1788 | ||
| GenBank mRNA | AK028712.1 | - | 13 | 1799 | ||
| AK031570.1 | - | 13 | 1789 | |||
| AK144239.1 | - | 13 | 1793 | |||