siRNA Details
| Gene Symbol: Rhof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175092.3 | NP_780301.1 | 5 | 1160 | ||
| GenBank mRNA | AK038837.1 | - | 928 | |||
| AK172212.1 | - | 5 | 928 | |||
| BC096597.1 | - | 5 | 950 | 4983233 | ||
| BC132355.1 | - | 992 | 40130632 | |||
| BC132357.1 | - | 992 | 40130634 | |||
| DV646753.1 | - | 2 | 582 | |||

Rhof
Genomic Map