siRNA Details
| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175443.5 | NP_780652.2 | 2 | 475 | ||
| GenBank mRNA | AK031665.1 | - | 2 | 261 | ||
| AK077179.1 | - | 2 | 214 | |||
| AK087818.1 | - | 1 | 273 | |||
| AY212244.1 | - | 2 | 66 | |||
| BC145351.1 | - | 2 | 254 | 9053907 | ||
| BC152310.1 | - | 2 | 413 | 40089976 | ||

Etnk2
Genomic Map