siRNA Details
| Gene Symbol: Mrgprb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175531.4 | NP_780740.2 | 2 | 961 | ||
| GenBank mRNA | AK029369.1 | - | 2 | 962 | ||
| BC107390.1 | - | 2 | 959 | 40057815 | ||
| BC107391.2 | - | 2 | 959 | 40057816 | ||
More Information
Set Membership: |
|

Mrgprb2
Genomic Map