siRNA Details
| Gene Symbol: Vkorc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178600.2 | NP_848715.1 | 3 | 500 | ||
| GenBank mRNA | AK002742.1 | - | 3 | 481 | ||
| AK003237.1 | - | 3 | 501 | |||
| AK008509.1 | - | 3 | 474 | |||
| AK008578.1 | - | 3 | 475 | |||
| AK013996.1 | - | 3 | 484 | |||
| BC031732.1 | - | 3 | 482 | 3991412 | ||

Vkorc1
Genomic Map