siRNA Details
| Gene Symbol: Zfp365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178679.2 | NP_848794.1 | 2 | 595 | ||
| GenBank mRNA | AJ516033.1 | - | 2 | 457 | ||
| AK031846.1 | - | 2 | 391 | |||
| AK033892.1 | - | 2 | 357 | |||
| AK077316.1 | - | 2 | 368 | |||
| BC080272.1 | - | 2 | 555 | 6837597 | ||
| CB519185.1 | - | 2 | 595 | |||

Zfp365
Genomic Map