siRNA Details
| Gene Symbol: Zfp365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178679.2 | NP_848794.1 | 5 | 1670 | ||
| GenBank mRNA | AJ516033.1 | - | 5 | 1532 | ||
| AK031846.1 | - | 5 | 1466 | |||
| AK033892.1 | - | 5 | 1432 | |||
| AK044655.1 | - | 4 | 753 | |||
| BC080272.1 | - | 5 | 1630 | 6837597 | ||

Zfp365
Genomic Map