siRNA Details
| Gene Symbol: Zfp592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178707.4 | NP_848822.2 | 3 | 927 | ||
| GenBank mRNA | AK042675.1 | - | 3 | 929 | ||
| AK129089.1 | - | 3 | 899 | |||
| AK143618.1 | - | 3 | 925 | |||
| BC059073.1 | - | 1 | 240 | |||
| BC094321.1 | - | 1 | 712 | 6401005 | ||
| BC118972.1 | - | 2 | 781 | 40098313 | ||
| BC118973.1 | - | 2 | 781 | 40098315 | ||

Zfp592
Genomic Map