siRNA Details
| Gene Symbol: Arrdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042591.1 | NP_001036056.1 | 1 | 386 | ||
| GenBank mRNA | AK084951.1 | - | 1 | 397 | ||
| AK088288.1 | - | 1 | 401 | |||
| AK122499.1 | - | 1 | 373 | |||
| BC044762.1 | - | 1 | 384 | |||
| BC054844.1 | - | 1 | 386 | 6493441 | ||
| DQ861994.1 | - | 1 | 131 | |||

Arrdc3
Genomic Map