siRNA Details
| Gene Symbol: Atad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_179203.3 | NP_849534.2 | 6 | 637 | ||
| GenBank mRNA | AF343079.1 | - | 6 | 683 | ||
| AK075921.1 | - | 6 | 633 | |||
| AK148974.1 | - | 6 | 636 | |||
| AK149702.1 | - | 6 | 636 | |||
| AK150037.1 | - | 6 | 636 | |||
| AK165724.1 | - | 6 | 635 | |||
| AK166099.1 | - | 6 | 633 | |||
| AK172038.1 | - | 6 | 633 | |||
| AK172350.1 | - | 6 | 638 | |||
| AK173141.1 | - | 6 | 585 | |||
| AK185628.1 | - | 6 | 366 | |||
| BC023301.1 | - | 6 | 574 | 3671787 | ||
| BC058373.1 | - | 6 | 621 | 6828037 | ||
| BC060036.1 | - | 6 | 569 | 6530155 | ||

Atad3a
Genomic Map