siRNA Details
| Gene Symbol: Clec18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181549.3 | NP_853527.2 | 1 | 189 | ||
| GenBank mRNA | AK132888.1 | - | 3 | 996 | ||
| AY223868.1 | - | 278 | ||||
| AY223869.1 | - | 249 | ||||
| AY223870.1 | - | 278 | ||||
| AY223871.1 | - | 278 | ||||
| AY223872.1 | - | 249 | ||||
| AY223873.1 | - | 249 | ||||

Clec18a
Genomic Map