siRNA Details
| Gene Symbol: Gadd45gip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183358.4 | NP_899202.3 | 2 | 689 | ||
| GenBank mRNA | AK002738.1 | - | 2 | 695 | ||
| AK003314.1 | - | 2 | 690 | |||
| AK009723.1 | - | 2 | 688 | |||
| AK019570.1 | - | 2 | 690 | |||
| AK028227.1 | - | 2 | 690 | |||
| BC022592.1 | - | 2 | 574 | |||
| BC061069.1 | - | 2 | 689 | 6740575 | ||

Gadd45gip1
Genomic Map