siRNA Details
| Gene Symbol: Gadd45gip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183358.4 | NP_899202.3 | 2 | 721 | ||
| GenBank mRNA | AK028227.1 | - | 2 | 722 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gadd45gip1| Gene Symbol: Gadd45gip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183358.4 | NP_899202.3 | 2 | 721 | ||
| GenBank mRNA | AK028227.1 | - | 2 | 722 | ||