siRNA Details
Gene Symbol: Scyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198021.2 | NP_932138.1 | 5 | 895 | ||
GenBank mRNA | AK139761.1 | - | 5 | 880 | ||
BC042443.1 | - | 5 | 895 | 4037878 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198021.2 | NP_932138.1 | 5 | 895 | ||
GenBank mRNA | AK139761.1 | - | 5 | 880 | ||
BC042443.1 | - | 5 | 895 | 4037878 |