siRNA Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198021.2 | NP_932138.1 | 16 | 2225 | ||
| GenBank mRNA | AK139747.1 | - | 8 | 811 | ||
| AK139761.1 | - | 16 | 2209 | |||
| BC042443.1 | - | 16 | 2225 | 4037878 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scyl2| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198021.2 | NP_932138.1 | 16 | 2225 | ||
| GenBank mRNA | AK139747.1 | - | 8 | 811 | ||
| AK139761.1 | - | 16 | 2209 | |||
| BC042443.1 | - | 16 | 2225 | 4037878 | ||