siRNA Details
| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198304.2 | NP_938046.2 | 4 | 201 | ||
| GenBank mRNA | AK052272.1 | - | 4 | 207 | ||
| AK129073.1 | - | 4 | 201 | |||
| BC028665.1 | - | 4 | 210 | |||
| BC032090.1 | - | 4 | 202 | |||
| BC096591.1 | - | 4 | 202 | 4916413 | ||

Nup188
Genomic Map