siRNA Details
| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198304.2 | NP_938046.2 | 9 | 737 | ||
| GenBank mRNA | AK052272.1 | - | 9 | 743 | ||
| AK129073.1 | - | 9 | 737 | |||
| BC032090.1 | - | 9 | 738 | |||
| BC096591.1 | - | 9 | 738 | 4916413 | ||

Nup188
Genomic Map