siRNA Details
| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198304.2 | NP_938046.2 | 44 | 5598 | ||
| GenBank mRNA | AK082836.1 | - | 11 | 1945 | ||
| AK129073.1 | - | 44 | 5598 | |||
| AK198439.1 | - | 1 | 192 | |||
| AK200842.1 | - | 1 | 191 | |||
| BC025526.1 | - | 11 | 1892 | |||
| BC032090.1 | - | 44 | 5600 | |||
| BC050199.1 | - | 9 | 1526 | |||
| BC056489.1 | - | 30 | 4177 | |||
| BC096591.1 | - | 44 | 5599 | 4916413 | ||

Nup188
Genomic Map