siRNA Details
| Gene Symbol: Slc22a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198650.2 | NP_941052.1 | 6 | 1027 | ||
| GenBank mRNA | AY194118.1 | - | 6 | 1026 | ||
| BC046588.1 | - | 6 | 1027 | 6309674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a20| Gene Symbol: Slc22a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198650.2 | NP_941052.1 | 6 | 1027 | ||
| GenBank mRNA | AY194118.1 | - | 6 | 1026 | ||
| BC046588.1 | - | 6 | 1027 | 6309674 | ||