siRNA Details
| Gene Symbol: Nsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198654.3 | NP_941056.3 | 1, 2 | 251 | ||
| GenBank mRNA | AK029421.1 | - | 1, 2 | 249 | ||
| BC029086.1 | - | 1, 2 | 241 | 3156934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsl1| Gene Symbol: Nsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198654.3 | NP_941056.3 | 1, 2 | 251 | ||
| GenBank mRNA | AK029421.1 | - | 1, 2 | 249 | ||
| BC029086.1 | - | 1, 2 | 241 | 3156934 | ||