siRNA Details
| Gene Symbol: Ccdc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199310.2 | NP_955014.2 | 4 | 925 | ||
| GenBank mRNA | AK132604.1 | - | 4 | 917 | ||
| AK133813.1 | - | 4 | 900 | |||
| AK147152.1 | - | 4 | 901 | |||
| AK152091.1 | - | 1 | 278 | |||
| BC023926.1 | - | 4 | 919 | 5324154 | ||
| BC052687.1 | - | 4 | 892 | |||

Ccdc32
Genomic Map