siRNA Details
| Gene Symbol: Gm9994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205249.1 | NP_001192178.1 | 1055 | |||
| GenBank mRNA | AY216522.1 | - | 1055 | |||
| Gene Symbol: Gal3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199366.4 | NP_955398.3 | 4 | 1311 | ||
| GenBank mRNA | AK144526.1 | - | 4 | 1310 | ||
| AK165692.1 | - | 4 | 1315 | |||
| BC132622.1 | - | 4 | 1090 | 40130899 | ||
| BC132624.1 | - | 4 | 1090 | 40130901 | ||

Gm9994
Genomic Map