siRNA Details
Gene Symbol: Chrm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205783.2 | NP_991352.2 | 1 | 910 | ||
GenBank mRNA | BC120615.1 | - | 1 | 910 | 40129538 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
