siRNA Details
| Gene Symbol: Chrm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205783.2 | NP_991352.2 | 1 | 1099 | ||
| GenBank mRNA | BC120615.1 | - | 1 | 1099 | 40129538 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrm5| Gene Symbol: Chrm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205783.2 | NP_991352.2 | 1 | 1099 | ||
| GenBank mRNA | BC120615.1 | - | 1 | 1099 | 40129538 | |
Set Membership: |
|