siRNA Details
| Gene Symbol: Kcnj4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008427.4 | NP_032453.3 | 2 | 1087 | ||
| GenBank mRNA | AB024304.1 | - | 1 | 972 | ||
| BC120593.1 | - | 2 | 1064 | 40129516 | ||
| BC137594.1 | - | 2 | 1064 | 8860613 | ||
| S71382.1 | - | 2 | 1088 | |||
| U11075.1 | - | 2 | 1057 | |||

Kcnj4
Genomic Map