siRNA Details
| Gene Symbol: Hoxc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010462.5 | NP_034592.2 | 2 | 1498 | ||
| GenBank mRNA | AK033186.1 | - | 2 | 1430 | ||
| AK138664.1 | - | 2 | 1488 | |||
| BC053405.1 | - | 2 | 1510 | 30059581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc10| Gene Symbol: Hoxc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010462.5 | NP_034592.2 | 2 | 1498 | ||
| GenBank mRNA | AK033186.1 | - | 2 | 1430 | ||
| AK138664.1 | - | 2 | 1488 | |||
| BC053405.1 | - | 2 | 1510 | 30059581 | ||