siRNA Details
| Gene Symbol: PSCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005672.4 | NP_005663.2 | 3 | 539 | ||
| NR_033343.1 | 3 | 692 | ||||
| GenBank mRNA | AF043498.1 | - | 3 | 501 | ||
| AJ297436.1 | - | 3 | 502 | |||
| AY358912.1 | - | 3 | 472 | |||
| BC023582.2 | - | 3 | 489 | 4840974 | ||
| BC048808.1 | - | 3 | 489 | |||
| BC065183.1 | - | 3 | 489 | 5557096 | ||
| CB993163.1 | - | 3 | 566 | |||

PSCA
Genomic Map