siRNA Details
| Gene Symbol: Zfp106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011743.2 | NP_035873.2 | 3 | 261 | ||
| GenBank mRNA | AK089359.1 | - | 3 | 259 | ||
| AK147887.1 | - | 3 | 262 | |||
| AK155019.1 | - | 3 | 259 | |||
| BC010440.1 | - | 2 | 101 | |||
| BC025424.1 | - | 3 | 250 | 4459713 | ||
| CJ104804.1 | - | 3 | 262 | |||

Zfp106
Genomic Map